• USA Home
  • 367087 - Poly(1,2-dihydro-2,2,4-trimethylquinoline)

EMAIL THIS PAGE TO A FRIEND
367087 Sigma-Aldrich

Poly(1,2-dihydro-2,2,4-trimethylquinoline)

technical grade

Purchase

Properties

grade   technical grade
transition temp   Tg (DSC) 78 °C (onset)

Description

Application

Antioxidant.

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
TQ2625000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?