• USA Home
  • 367214 - 3-(Bromomethyl)-2,4,10-trioxaadamantane

EMAIL THIS PAGE TO A FRIEND
367214 Sigma-Aldrich

3-(Bromomethyl)-2,4,10-trioxaadamantane

96%

Synonym: 3-(Bromomethyl)-2,4,10-trioxatricyclo[3.3.1.13.7]decane, Bromo(2,3,10-trioxatricyclo[3.3.1.13.7]dec-3-yl)methane

  • CAS Number 157371-80-7

  • Empirical Formula (Hill Notation) C8H11BrO3

  • Molecular Weight 235.08

  •  MDL number MFCD00077628

Purchase

Properties

assay   96%
mp   94-96 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

17624 3-(Bromomethyl)-2,4,10-trioxatricyclo[3.3.1.13.7]decane, ≥97.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?