• USA Home
  • 367370 - (±)-2-Amino-4-phosphonobutyric acid

EMAIL THIS PAGE TO A FRIEND
367370 Aldrich

(±)-2-Amino-4-phosphonobutyric acid

97%

Synonym: (±)-AP-4

  • CAS Number 20263-07-4

  • Empirical Formula (Hill Notation) C4H10NO5P

  • Molecular Weight 183.10

  •  MDL number MFCD00013999

Purchase

Properties

assay   97%
SMILES string   NC(CCP(O)(O)=O)C(O)=O
InChI   1S/C4H10NO5P/c5-3(4(6)7)1-2-11(8,9)10/h3H,1-2,5H2,(H,6,7)(H2,8,9,10)
InChI key   DDOQBQRIEWHWBT-UHFFFAOYSA-N

Description

Biochem/physiol Actions

NMDA glutamate receptor antagonist.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?