• USA Home
  • 367761 - Cesium trifluoroacetate

EMAIL THIS PAGE TO A FRIEND
367761 Aldrich

Cesium trifluoroacetate

≥99.99%

Synonym: CsTFA, Trifluoroacetic acid cesium salt

  • CAS Number 21907-50-6

  • Linear Formula CF3COOCs

  • Molecular Weight 245.92

  •  Beilstein Registry Number 4161420

  •  MDL number MFCD00051304

Purchase

Properties

InChI Key   RJYSYRSELCQCSO-UHFFFAOYSA-M
assay   ≥99.99%
mp   114-116 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?