• USA Home
  • 367788 - Purpurogallin tetramethyl ether

EMAIL THIS PAGE TO A FRIEND
367788 Sigma-Aldrich

Purpurogallin tetramethyl ether

99%

  • CAS Number 6273-57-0

  • Empirical Formula (Hill Notation) C15H16O5

  • Molecular Weight 276.28

  •  MDL number MFCD00075499

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?