• USA Home
  • 368431 - D-α,β-Cyclohexylideneglycerol

EMAIL THIS PAGE TO A FRIEND
368431 Sigma-Aldrich

D-α,β-Cyclohexylideneglycerol

98%

Synonym: (S)-(+)-1,4-Dioxaspiro[4.5]decane-2-methanol

  • CAS Number 95335-91-4

  • Empirical Formula (Hill Notation) C9H16O3

  • Molecular Weight 172.22

  •  Beilstein/REAXYS Number 108000

  •  MDL number MFCD08276891

Purchase

Properties

InChI Key   XUGYZVQSZWPABZ-QMMMGPOBSA-N
assay   98%
optical activity   [α]20/D +9.5°, neat
refractive index   n20/D 1.477 (lit.)
bp   118-120 °C/5 mmHg (lit.)
density   1.096 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

29400 D-α,β-Cyclohexylideneglycerol, ≥98.0% (sum of enantiomers, GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?