• USA Home
  • 368504 - 3-Fluoro-1,2-propanediol

EMAIL THIS PAGE TO A FRIEND
368504 Aldrich

3-Fluoro-1,2-propanediol

98%

Synonym: α-Fluorohydrin

  • CAS Number 453-16-7

  • Linear Formula FCH2CH(OH)CH2OH

  • Molecular Weight 94.08

  •  MDL number MFCD00075515

Purchase

Properties

InChI Key   PQDNJBQKAXAXBQ-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.422 (lit.)
bp   102-103 °C/13 mmHg(lit.)
density   1.1 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
RTECS 
TY6791500
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?