• USA Home
  • 368687 - 2′-Chloroethyl β-D-fructopyranoside

EMAIL THIS PAGE TO A FRIEND
368687 Sigma-Aldrich

2′-Chloroethyl β-D-fructopyranoside

98%

  • CAS Number 84543-36-2

  • Empirical Formula (Hill Notation) C8H15ClO6

  • Molecular Weight 242.65

  •  MDL number MFCD00075519

Purchase

Properties

assay   98%
optical activity   [α]23/D −146°, c = 1 in H2O
mp   139-141 °C (dec.) (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?