• USA Home
  • 368695 - D-Lactitol monohydrate

EMAIL THIS PAGE TO A FRIEND
368695 Aldrich

D-Lactitol monohydrate

98%

Synonym: 4-O-β-D-Galactopyranosyl-D-glucitol

  • CAS Number 81025-04-9

  • Empirical Formula (Hill Notation) C12H24O11 · H2O

  • Molecular Weight 362.33

  •  Beilstein Registry Number 4218679

  •  EC Number 209-566-5

  •  MDL number MFCD00150767

Purchase

Properties

InChI Key   LXMBXZRLTPSWCR-XBLONOLSSA-N
assay   98%
optical activity   [α]20/D +13°, c = 1 in H2O
mp   95-98 °C (lit.)

Description

Biochem/physiol Actions

Sugar alcohol sweet tastant detectable by humans.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

L3520 D-Lactitol monohydrate, ~99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?