• USA Home
  • 370126 - N,O-Bis(4-methoxybenzyloxycarbonyl)-(S)-2-amino-3-(4-hydroxyphenyl)propionitrile

EMAIL THIS PAGE TO A FRIEND
370126 Aldrich

N,O-Bis(4-methoxybenzyloxycarbonyl)-(S)-2-amino-3-(4-hydroxyphenyl)propionitrile

96%

  • CAS Number 73148-72-8

  • Linear Formula CH3OC6H4CH2OCO2C6H4CH2CH(NHCO2CH2C6H4OCH3)CN

  • Molecular Weight 490.50

  •  MDL number MFCD00075549

  •  PubChem Substance ID 24863058

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Cyanides/Nitriles, Organic Building Blocks More...
InChI Key   IEWDJSXKLMDHDC-QFIPXVFZSA-N
assay   96%
mp   108-112 °C(lit.)

Description

Packaging

100 mg in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?