• USA Home
  • 37034 - (3R,4R)-(−)-1-Benzyl-3,4-pyrrolidinediol

EMAIL THIS PAGE TO A FRIEND
37034 Sigma-Aldrich

(3R,4R)-(−)-1-Benzyl-3,4-pyrrolidinediol

≥97.0% (sum of enantiomers, HPLC)

  • CAS Number 163439-82-5

  • Empirical Formula (Hill Notation) C11H15NO2

  • Molecular Weight 193.24

  •  Beilstein/REAXYS Number 6478637

  •  MDL number MFCD01073894

  •  PubChem Substance ID 57649705

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Pyrrolidines More...
InChI Key   QJRIUWQPJVPYSO-GHMZBOCLSA-N
assay   ≥97.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D −33.6±3°, c = 1.05% in methanol

Description

General description

(3R,4R)-1-benzyl-3,4-bis(benzyloxy)pyrrolidinediol, a chiral pyrrolidine derivative, is a chiral building block for auxiliaries and biologically active compounds.

Application

(3R,4R)-1-benzyl-3,4-bis(benzyloxy)pyrrolidinediol may be used in the synthesis of pyrrolidine based iminocyclitol inhibitors, (3R,4R)-1-benzyl-3,4-bis(benzyloxy)pyrrolidine and (3R,4R)-pyrrolidine-3,4-diol, respectively.

Packaging

500 mg in poly tube

Other Notes

Chiral building block

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?