• USA Home
  • 371106 - 1-Deoxy-1-(methylamino)-D-galactitol

EMAIL THIS PAGE TO A FRIEND
371106 Sigma-Aldrich

1-Deoxy-1-(methylamino)-D-galactitol

99%

  • CAS Number 7115-46-0

  • Empirical Formula (Hill Notation) C7H17NO5

  • Molecular Weight 195.21

  •  MDL number MFCD00134351

  •  PubChem Substance ID 24863091

Purchase

Properties

assay   99%
optical activity   [α]20/D −16°, c = 1 in methanol
mp   133-136 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?