• USA Home
  • 371157 - (1S,2S)-(−)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]-1,2-ethanediamine

EMAIL THIS PAGE TO A FRIEND
371157 Sigma-Aldrich

(1S,2S)-(−)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]-1,2-ethanediamine

99%

  • CAS Number 205873-26-3

  • Empirical Formula (Hill Notation) C18H18F6N2

  • Molecular Weight 376.34

  •  MDL number MFCD00075561

Purchase

Properties

assay   99%
optical activity   [α]20/D −15°, c = 0.5 in chloroform
mp   113-115 °C (lit.)
  114-118 °C (lit.)
SMILES string   CN[C@H]([C@@H](NC)c1cccc(c1)C(F)(F)F)c2cccc(c2)C(F)(F)F
InChI   1S/C18H18F6N2/c1-25-15(11-5-3-7-13(9-11)17(19,20)21)16(26-2)12-6-4-8-14(10-12)18(22,23)24/h3-10,15-16,25-26H,1-2H3/t15-,16-/m0/s1
InChI key   SBGOGHODWUZHIY-HOTGVXAUSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?