• USA Home
  • 371556 - N-[4-(2,4-Di-tert-pentylphenoxy)butyl]-1-hydroxy-2-naphthalenecarboxamide

EMAIL THIS PAGE TO A FRIEND
371556

N-[4-(2,4-Di-tert-pentylphenoxy)butyl]-1-hydroxy-2-naphthalenecarboxamide

99%

Purchase

Properties

assay   99%
SMILES string   CCC(C)(C)c1ccc(OCCCCNC(=O)c2ccc3ccccc3c2O)c(c1)C(C)(C)CC
InChI   1S/C31H41NO3/c1-7-30(3,4)23-16-18-27(26(21-23)31(5,6)8-2)35-20-12-11-19-32-29(34)25-17-15-22-13-9-10-14-24(22)28(25)33/h9-10,13-18,21,33H,7-8,11-12,19-20H2,1-6H3,(H,32,34)
InChI key   DHEJKONKJWLHGP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?