• USA Home
  • 371793 - 5-(3-Amino-5-oxo-2-pyrazolin-1-yl)-2-phenoxybenzenesulfonic acid

EMAIL THIS PAGE TO A FRIEND
371793 Sigma-Aldrich

5-(3-Amino-5-oxo-2-pyrazolin-1-yl)-2-phenoxybenzenesulfonic acid

technical grade, 92%

  • CAS Number 30479-81-3

  • Empirical Formula (Hill Notation) C15H13N3O5S

  • Molecular Weight 347.35

  •  MDL number MFCD00134368

  •  PubChem Substance ID 24863130

Purchase

Properties

Related Categories Building Blocks, C12 to C37, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazoles More...
grade   technical grade
assay   92%
mp   297 °C (dec.) (lit.)
SMILES string   NC1=NN(C(=O)C1)c2ccc(Oc3ccccc3)c(c2)S(O)(=O)=O
InChI   1S/C15H13N3O5S/c16-14-9-15(19)18(17-14)10-6-7-12(13(8-10)24(20,21)22)23-11-4-2-1-3-5-11/h1-8H,9H2,(H2,16,17)(H,20,21,22)
InChI key   ZZSAILZSWKPTCT-UHFFFAOYSA-N

Description

General description

5-(3-Amino-5-oxo-2-pyrazolin-1-yl)-2-phenoxybenzenesulfonic acid is a heterocyclic building block.

Packaging

5, 25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?