• USA Home
  • 371920 - 2,2,3,3-Tetrafluoropropyl acrylate

EMAIL THIS PAGE TO A FRIEND
371920 Sigma-Aldrich

2,2,3,3-Tetrafluoropropyl acrylate

96%

Purchase

Properties

assay   96%
contains   hydroquinone as stabilizer
refractive index   n20/D 1.365 (lit.)
bp   132-133 °C(lit.)
density   1.317 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
UN 3272 3/PG 3
WGK Germany 
3
Flash Point(F) 
114 °F
Flash Point(C) 
46 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?