• USA Home
  • 371947 - Allyl 1,1,2,2-tetrafluoroethyl ether

EMAIL THIS PAGE TO A FRIEND
371947

Allyl 1,1,2,2-tetrafluoroethyl ether

99%

Purchase

Properties

assay   99%
SMILES string   FC(F)C(F)(F)OCC=C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3271 3 / PGII
WGK Germany 
3
Flash Point(F) 
23 °F
Flash Point(C) 
-5 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?