• USA Home
  • 372498 - Borobetaine methyl ester

EMAIL THIS PAGE TO A FRIEND
372498 Aldrich

Borobetaine methyl ester

98%

Synonym: (Methoxycarbonyl)borane trimethylamine complex, Trimethylamine-carbomethoxyborane

Purchase

Properties

Related Categories Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents, Other Boronic Acids and Derivatives
InChI Key   RIFGGBKCDHRFFC-UHFFFAOYSA-N
assay   98%
mp   92-93 °C(lit.)

Description

Legal Information

Boro... is a trademark of Boron Biologicals, Inc.

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
ED7685000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?