• USA Home
  • 373745 - 3-(2-Hydroxyethylamino)-5,5-dimethyl-2-cyclohexen-1-one

EMAIL THIS PAGE TO A FRIEND
373745 Sigma-Aldrich

3-(2-Hydroxyethylamino)-5,5-dimethyl-2-cyclohexen-1-one

98%

Purchase

Properties

assay   98%
mp   91-93 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?