• USA Home
  • 374792 - (−)-Bis[(S)-1-(ethoxycarbonyl)ethyl] fumarate

EMAIL THIS PAGE TO A FRIEND
374792 Sigma-Aldrich

(−)-Bis[(S)-1-(ethoxycarbonyl)ethyl] fumarate

97%

Synonym: (−)-Fumaric acid bis[(S)-1-(ethoxycarbonyl)ethyl] ester

  • CAS Number 111293-23-3

  • Linear Formula [=CHCO2CH(CH3)CO2C2H5]2

  • Molecular Weight 316.30

  •  Beilstein/REAXYS Number 4201513

  •  MDL number MFCD00075600

  •  PubChem Substance ID 24863337

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters, Organic Building Blocks More...
InChI Key   PVSLHMRMQFZKMW-GOPXOUGQSA-N
assay   97%
optical activity   [α]20/D −8°, c = 0.5 in chloroform
bp   151 °C/0.4 mmHg(lit.)
density   1.139 g/mL at 20 °C (lit.)

Description

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?