• USA Home
  • 374814 - 4-Fluorophenyl chlorothionoformate

EMAIL THIS PAGE TO A FRIEND
374814 Sigma-Aldrich

4-Fluorophenyl chlorothionoformate

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.558 (lit.)
bp   94-95 °C/10 mmHg (lit.)
density   1.363 g/mL at 25 °C (lit.)
SMILES string   Fc1ccc(OC(Cl)=S)cc1
InChI   1S/C7H4ClFOS/c8-7(11)10-6-3-1-5(9)2-4-6/h1-4H
InChI key   HZBXSUFBKXPOAF-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGIII
WGK Germany 
3
Flash Point(F) 
183.2 °F
Flash Point(C) 
84 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?