• USA Home
  • 374911 - (S)-(+)-2-(Anilinomethyl)pyrrolidine

EMAIL THIS PAGE TO A FRIEND
374911 Sigma-Aldrich

(S)-(+)-2-(Anilinomethyl)pyrrolidine

96%

  • CAS Number 64030-44-0

  • Empirical Formula (Hill Notation) C11H16N2

  • Molecular Weight 176.26

  •  Beilstein/REAXYS Number 472065

  •  MDL number MFCD00015890

  •  PubChem Substance ID 24863349

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Proline-Based Organocatalysts
InChI Key   MCHWKJRTMPIHRA-NSHDSACASA-N
assay   96%
optical activity   [α]20/D +19°, c = 1 in ethanol
bp   111-112 °C/0.55 mmHg(lit.)
density   1.046 g/mL at 25 °C (lit.)

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3267 8 / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Chiral Vicinal Diamines for Asymmetric Synthesis

Chiral vicinal diamines are of tremendous interest to the synthetic chemist as they are found in many chiral catalysts and pharmaceuticals. Currently, there is no unified approach to making these chi...
ChemFiles 2007, 7.9, 7.
Keywords: Ligands, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?