• USA Home
  • 375306 - 1-(2-Fluorophenyl)piperazine monohydrochloride

EMAIL THIS PAGE TO A FRIEND
375306 Sigma-Aldrich

1-(2-Fluorophenyl)piperazine monohydrochloride

98%

  • CAS Number 1011-16-1

  • Empirical Formula (Hill Notation) C10H13FN2 · HCl

  • Molecular Weight 216.68

  •  MDL number MFCD00134409

  •  PubChem Substance ID 24863380

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   JVQOFQHGEFLRRQ-UHFFFAOYSA-N
assay   98%
mp   187-189 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?