• USA Home
  • 375489 - 3,4,5-Trichlorobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
375489 Sigma-Aldrich

3,4,5-Trichlorobenzotrifluoride

97%

Purchase

Properties

assay   97%
refractive index   n20/D 1.5 (lit.)
bp   200-202 °C (lit.)
density   1.6 g/mL at 25 °C (lit.)
SMILES string   FC(F)(F)c1cc(Cl)c(Cl)c(Cl)c1
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?