• USA Home
  • 375586 - Barium nitrite hydrate

EMAIL THIS PAGE TO A FRIEND
375586 Sigma-Aldrich

Barium nitrite hydrate

technical grade, 90%

Purchase

Properties

grade   technical grade
assay   90%
impurities   7-8% KClO4
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2627 - class 5.1 - PG 2 - EHS - Nitrites, inorganic, n.o.s., HI: all (not BR)
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?