• USA Home
  • 375780 - 3,5-Dinitro-L-tyrosine monohydrate

EMAIL THIS PAGE TO A FRIEND
375780 Sigma-Aldrich

3,5-Dinitro-L-tyrosine monohydrate

97%

  • CAS Number 71876-88-5

  • Linear Formula (O2N)2C6H2-4-(OH)CH2CH(NH2)CO2H·H2O

  • Molecular Weight 289.20

  •  MDL number MFCD00150534

Purchase

Properties

assay   97%
optical activity   [α]20/D +12°, c = 1 in 1 M HCl
mp   220 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?