• USA Home
  • 376272 - Triethoxy(3-thiocyanatopropyl)silane

EMAIL THIS PAGE TO A FRIEND
376272 Sigma-Aldrich

Triethoxy(3-thiocyanatopropyl)silane

95%

Synonym: (3-Thiocyanatopropyl)triethoxysilane

  • CAS Number 34708-08-2

  • Empirical Formula (Hill Notation) C10H21NO3SSi

  • Molecular Weight 263.43

  •  Beilstein/REAXYS Number 1949834

  •  MDL number MFCD00043235

Purchase

Properties

assay   95%
density   1.031 g/mL at 20 °C (lit.)
SMILES string   CCO[Si](CCCSC#N)(OCC)OCC
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?