• USA Home
  • 376884 - 2-Amino-5-bromo-3-nitropyridine

EMAIL THIS PAGE TO A FRIEND
376884 Sigma-Aldrich

2-Amino-5-bromo-3-nitropyridine

97%

  • CAS Number 6945-68-2

  • Empirical Formula (Hill Notation) C5H4BrN3O2

  • Molecular Weight 218.01

  •  MDL number MFCD00047441

  •  PubChem Substance ID 24863473

Purchase

Properties

Related Categories Building Blocks, C5, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
InChI Key   QOOCOFOGYRQPPN-UHFFFAOYSA-N
assay   97%
mp   205-208 °C (lit.)

Description

Packaging

1, 5 g in glass bottle

Application

2-Amino-5-bromo-3-nitropyridine may be used in the preparation of azaquinoxalinedione.

General description

2-Amino-5-bromo-3-nitropyridine is formed as intermediate in the synthesis of 2,3-diaminopyridine.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?