• USA Home
  • 377503 - Isobutyltriphenylphosphonium bromide

EMAIL THIS PAGE TO A FRIEND
377503 Sigma-Aldrich

Isobutyltriphenylphosphonium bromide

97%

Purchase

Properties

assay   97%
mp   200-202 °C (lit.)

Description

Application

Used to prepare the novel sponge-derived amino acid bengamide E and "hot pepper"capsaicinoids.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?