EMAIL THIS PAGE TO A FRIEND
378798 Aldrich

Barium fluoride

optical grade, random crystals, ≥99.9%

Purchase

Properties

grade   optical grade
InChI Key   OYLGJCQECKOTOL-UHFFFAOYSA-L
assay   ≥99.9%
form   random crystals
bp   2260 °C(lit.)
density   4.89 g/mL at 25 °C(lit.)

Description

Features and Benefits

Primary glass modifier in fluorozirconate glasses.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1564 6.1 / PGIII
WGK Germany 
1
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?