• USA Home
  • 379360 - 6′-Methoxy-2′-propiononaphthone

EMAIL THIS PAGE TO A FRIEND
379360 Sigma-Aldrich

6′-Methoxy-2′-propiononaphthone

98%

Purchase

Properties

Related Categories Building Blocks, C13 to C14, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   LWOTXBQKJLOAOZ-UHFFFAOYSA-N
assay   98%
bp   212-215 °C/14 mmHg (lit.)
mp   110-112 °C (lit.)

Description

General description

6′−Methoxy-2′−propiononaphthone is a building block useful in organic synthesis. Its density has been reported.

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?