EMAIL THIS PAGE TO A FRIEND
379654 Sigma-Aldrich

[5,6]-Fullerene-C70

Green Alternative

95%

Synonym: Buckminsterfullerene C70, Fullerene-C70

Purchase

Description

Other Notes

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Fullerenes for Bioscience & Photovoltaic Applications

Twenty-five years ago, it was discovered that carbon vapor preferentially condenses into an exceedingly stable cluster with 60 carbon atoms (C60), and it was hypothesized that the 60 atoms were arran...
Michael D. Diener

TDA Research
12345 W52nd Ave, Wheat Ridge, CO 80033
Keywords: AGE, Analytical Chemistry, Antivirals, Combustion, Cosmetics, Electronics, Magnetic resonance imaging, Mass spectrometry, Nanomaterials, Organic electronics, Photovoltaics, Polymerization reactions, Reductions, Semiconductor, Solar cells, Solvents, Transfection, Vitamins

Soluble Pentacene Precursors

In order to introduce un-functionalized pentacene within a device such as an organic field effect transistor (OFET), soluble pentacene precursors, which readily transform into pentacene upon heating ...
Prof. Cherie R. Kagan

Associate Professor, Department of Materials Science and Engineering,
University of Pennsylvania, Philadelphia, PA 19104
Keywords: Deposition, Evaporation, Semiconductor, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?