• USA Home
  • 380148 - (4S,5R)-(+)-1,5-Dimethyl-4-phenyl-2-imidazolidinone

EMAIL THIS PAGE TO A FRIEND
380148 Sigma-Aldrich

(4S,5R)-(+)-1,5-Dimethyl-4-phenyl-2-imidazolidinone

97%

  • CAS Number 112791-04-5

  • Empirical Formula (Hill Notation) C11H14N2O

  • Molecular Weight 190.24

  •  Beilstein/REAXYS Number 4905832

  •  MDL number MFCD00066231

  •  PubChem Substance ID 24863723

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Imidazolines/Imidazolidines More...
assay   97%
optical activity   [α]20/D +45°, c = 1 in methanol
mp   173-176 °C (lit.)
SMILES string   C[C@@H]1[C@@H](NC(=O)N1C)c2ccccc2
InChI   1S/C11H14N2O/c1-8-10(12-11(14)13(8)2)9-6-4-3-5-7-9/h3-8,10H,1-2H3,(H,12,14)/t8-,10-/m1/s1
InChI key   BGPAZBKCZVVZGF-PSASIEDQSA-N

Description

Application

Employed in the synthesis of optically active mono-, di-, and trihydroxy compounds.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?