• USA Home
  • 380504 - 3-Chloro-4-fluorotoluene

EMAIL THIS PAGE TO A FRIEND
380504 Aldrich

3-Chloro-4-fluorotoluene

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.501(lit.)
bp   167-169 °C(lit.)
density   1.182 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
131 °F
Flash Point(C) 
55 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?