• USA Home
  • 380571 - N-Acetonylphthalimide

EMAIL THIS PAGE TO A FRIEND
380571 Sigma-Aldrich

N-Acetonylphthalimide

98%

  • CAS Number 3416-57-7

  • Empirical Formula (Hill Notation) C11H9NO3

  • Molecular Weight 203.19

  •  MDL number MFCD00088346

Purchase

Properties

assay   98%
SMILES string   CC(=O)CN1C(=O)c2ccccc2C1=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?