• USA Home
  • 381217 - 2′-Bromo-4′-fluoroacetanilide

EMAIL THIS PAGE TO A FRIEND
381217 Sigma-Aldrich

2′-Bromo-4′-fluoroacetanilide

99%

Purchase

Properties

assay   99%
mp   117-119 °C (lit.)

Description

General description

2′-Bromo-4′-fluoroacetanilide is an organic building block.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?