EMAIL THIS PAGE TO A FRIEND
383031 Sigma-Aldrich

Niobium(V) oxide

lumps, 99.8% trace metals basis

Synonym: Niobium pentoxide

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science, Niobium,
InChI Key   ZKATWMILCYLAPD-UHFFFAOYSA-N
assay   99.8% trace metals basis
form   lumps
density   4.47 g/mL at 25 °C (lit.)

Description

Application

Promising results were obtained using niobium(V) oxide as an alternate electrode to lithium metal in advanced fuel cells.

Features and Benefits

Applied in the solid state formation of an unusual cation deficient perovskite, Ba7Nb4MoO20.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
QU0500000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?