EMAIL THIS PAGE TO A FRIEND
383740 Sigma-Aldrich

2,4-Dimethylphenol

technical grade, 90%

Synonym: 4-Hydroxy-m-xylene, asym.-m-Xylenol

  • CAS Number 105-67-9

  • Linear Formula (CH3)2C6H3OH

  • Molecular Weight 122.16

  •  Beilstein/REAXYS Number 636244

  •  EC Number 203-321-6

  •  MDL number MFCD00002233

Purchase

Properties

grade   technical grade
vapor pressure   0.1 mmHg ( 25 °C)
InChI Key   KUFFULVDNCHOFZ-UHFFFAOYSA-N
assay   90%
refractive index   n20/D 1.538 (lit.)
bp   211-212 °C(lit.)
mp   22-23 °C (lit.)
density   1.011 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2261 6.1 / PGII
WGK Germany 
3
RTECS 
ZE5600000
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?