• USA Home
  • 38425 - 2-(Diisopropylamino)ethanol

EMAIL THIS PAGE TO A FRIEND
38425 Sigma-Aldrich

2-(Diisopropylamino)ethanol

purum, ≥98.0% (GC)

Synonym: N,N-Diisopropylethanolamine

  • CAS Number 96-80-0

  • Linear Formula [(CH3)2CH]2NCH2CH2OH

  • Molecular Weight 145.24

  •  Beilstein/REAXYS Number 1697955

  •  EC Number 202-536-2

  •  MDL number MFCD00008869

  •  PubChem Substance ID 329755761

Purchase

Properties

Related Categories Amino Alcohols, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
vapor density   5 (vs air)
vapor pressure   <1 mmHg ( 20 °C)
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.442 (lit.)
  n20/D 1.442
bp   187-192 °C (lit.)
density   0.826 g/mL at 25 °C (lit.)
SMILES string   CC(C)N(CCO)C(C)C
InChI   1S/C8H19NO/c1-7(2)9(5-6-10)8(3)4/h7-8,10H,5-6H2,1-4H3
InChI key   ZYWUVGFIXPNBDL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
2
RTECS 
KK5950000
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?