• USA Home
  • 38481 - 2,3-Dilauroyl-sn-glycerol

EMAIL THIS PAGE TO A FRIEND
38481 Sigma-Aldrich

2,3-Dilauroyl-sn-glycerol

≥99.0% (TLC)

Synonym: (R)-1,2-Dilaurin, (R)-Glycerol 1,2-dilaurate

  • CAS Number 61475-84-1

  • Empirical Formula (Hill Notation) C27H52O5

  • Molecular Weight 456.70

  •  Beilstein/REAXYS Number 1807483

  •  MDL number MFCD00674509

Purchase

Properties

assay   ≥99.0% (TLC)
optical activity   [α]20/D +3.5±1°, c = 3% in chloroform
mp   45-49 °C (lit.)
SMILES string   CCCCCCCCCCCC(=O)OC[C@@H](CO)OC(=O)CCCCCCCCCCC
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?