EMAIL THIS PAGE TO A FRIEND
38488 Aldrich

Dilongifolylborane

≥98.0% (CH)

Purchase

Properties

assay   ≥98.0% (CH)
mp   169-172 °C

Description

Other Notes

For the asymmetric hydroboration especially of aliphatic, trisubstituted, acyclic olefins

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?