• USA Home
  • 385298 - (2S)-(−)-2,2′-Oxybis(octahydro-7,8,8-trimethyl-4,7-methanobenzofuran)

EMAIL THIS PAGE TO A FRIEND
385298 Sigma-Aldrich

(2S)-(−)-2,2′-Oxybis(octahydro-7,8,8-trimethyl-4,7-methanobenzofuran)

99%

Purchase

Properties

Related Categories Asymmetric Synthesis, Benzofurans, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks More...
InChI Key   PMKIEOMARWISLE-HDFUADCPSA-N
assay   99%
optical activity   [α]20/D −172°, c = 0.8 in methylene chloride
mp   149-151 °C (lit.)

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?