• USA Home
  • 385719 - 4,5-Diphenyl-2-oxazolethiol

EMAIL THIS PAGE TO A FRIEND
385719 Sigma-Aldrich

4,5-Diphenyl-2-oxazolethiol

98%

Purchase

Properties

assay   98%
mp   254-257 °C (lit.)
SMILES string   Sc1nc(-c2ccccc2)c(o1)-c3ccccc3
InChI   1S/C15H11NOS/c18-15-16-13(11-7-3-1-4-8-11)14(17-15)12-9-5-2-6-10-12/h1-10H,(H,16,18)
InChI key   NYQSTRUSYDZNHC-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?