• USA Home
  • 385735 - 5-(Octadecyloxy)isophthalic acid

EMAIL THIS PAGE TO A FRIEND
385735 Sigma-Aldrich

5-(Octadecyloxy)isophthalic acid

98%

Purchase

Properties

Related Categories Carboxylic Acid Monomers, Materials Science, Monomers, Polymer Science
assay   98%
mp   157-161 °C (lit.)
SMILES string   CCCCCCCCCCCCCCCCCCOc1cc(cc(c1)C(O)=O)C(O)=O
InChI   1S/C26H42O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-31-24-20-22(25(27)28)19-23(21-24)26(29)30/h19-21H,2-18H2,1H3,(H,27,28)(H,29,30)
InChI key   JAJBICIOMQKJMS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?