• USA Home
  • 385875 - Poly[2,6-bis(hydroxymethyl)-4-methylphenol-co-4-hydroxybenzoic acid]

EMAIL THIS PAGE TO A FRIEND
385875 Sigma-Aldrich

Poly[2,6-bis(hydroxymethyl)-4-methylphenol-co-4-hydroxybenzoic acid]

Purchase

Properties

mp   197 °C (dec.) (lit.)
SMILES string   [H]C([H])=O.Cc1ccc(O)cc1.OC(=O)c2ccc(O)cc2
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?