• USA Home
  • 385948 - 4-Hydroxy-3-(morpholinomethyl)benzoic acid hydrate

EMAIL THIS PAGE TO A FRIEND
385948 Sigma-Aldrich

4-Hydroxy-3-(morpholinomethyl)benzoic acid hydrate

90%, technical grade

  • CAS Number 143269-99-2

  • Empirical Formula (Hill Notation) C12H15NO4 · xH2O

  • Molecular Weight 237.25 (anhydrous basis)

  •  MDL number MFCD00192218

  •  PubChem Substance ID 24864060

Purchase

Properties

grade   technical grade
assay   90%
mp   168 °C (dec.) (lit.)
SMILES string   [H]O[H].OC(=O)c1ccc(O)c(CN2CCOCC2)c1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?