• USA Home
  • 386081 - 2-Methyl-5-phenyl-3-(3-sulfopropyl)benzoxazolium hydroxide inner salt

EMAIL THIS PAGE TO A FRIEND
386081 Sigma-Aldrich

2-Methyl-5-phenyl-3-(3-sulfopropyl)benzoxazolium hydroxide inner salt

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Oxazoles
mp   286-288 °C (lit.)
SMILES string   Cc1oc2ccc(cc2[n+]1CCCS([O-])(=O)=O)-c3ccccc3
InChI   1S/C17H17NO4S/c1-13-18(10-5-11-23(19,20)21)16-12-15(8-9-17(16)22-13)14-6-3-2-4-7-14/h2-4,6-9,12H,5,10-11H2,1H3
InChI key   DKMBQSTXNOURJD-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?