EMAIL THIS PAGE TO A FRIEND
386340 Sigma-Aldrich

4,4′-DDT

98%

Synonym: 1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane, 1,1-Bis(4-chlorophenyl)-2,2,2-trichloroethane

  • CAS Number 50-29-3

  • Linear Formula (ClC6H4)2CHCCl3

  • Molecular Weight 354.49

  •  Beilstein/REAXYS Number 1882657

  •  EC Number 200-024-3

  •  MDL number MFCD00000802

  •  PubChem Substance ID 24864083

Purchase

Properties

Related Categories Aryl, Building Blocks, C13 to C37+, Chemical Synthesis, Halogenated Hydrocarbons,
assay   98%
mp   107-110 °C (lit.)
SMILES string   Clc1ccc(cc1)C(c2ccc(Cl)cc2)C(Cl)(Cl)Cl
InChI   1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H
InChI key   YVGGHNCTFXOJCH-UHFFFAOYSA-N
Gene Information   mouse ... Esr1(13982)
rat ... Ar(24208)

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - SMP - RQ - Toxic solid, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
KJ3325000
Flash Point(F) 
161.6 - 170.6 °F
Flash Point(C) 
72 - 77 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Related Content

Carcinogen Selector - Hematopoietic System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Lung

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Urinary System - Kidney

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Urinary System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?