• USA Home
  • 386677 - 7,10-Dimethylbenz[c]acridine

EMAIL THIS PAGE TO A FRIEND
386677 Sigma-Aldrich

7,10-Dimethylbenz[c]acridine

technical grade

  • CAS Number 2381-40-0

  • Empirical Formula (Hill Notation) C19H15N

  • Molecular Weight 257.33

  •  MDL number MFCD00056821

  •  PubChem Substance ID 24864110

Purchase

Properties

Related Categories Acridines, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
grade   technical grade
InChI Key   RCWNHXIFXQTICS-UHFFFAOYSA-N
mp   147-149 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CU3485000

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?