• USA Home
  • 386979 - Poly-D-lysine hydrobromide

EMAIL THIS PAGE TO A FRIEND
386979 Aldrich

Poly-D-lysine hydrobromide

average Mv 1,000-4,000

  • CAS Number 27964-99-4

  • Linear Formula D-Lys-(D-Lys)/n-D-Lys · xHBr

Purchase

Properties

mol wt   average Mv 1,000-4,000
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?